Imprimer
Air admittance valve
Fournisseur: Vacuubrand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
183-1201EA 166 EUR
183-1201
Air admittance valve
Vannes Vannes de régulation d'air
These valves are compact air admittance valves with small flange. The VB valve is manually operated. On rotating the knurled screw cap the valve disc is lifted off the seating and air enters through the holes. The valve disc is rotatable. Therefore the valve seat seal is prevented against wear during opening and closing.
Leakage rate: 1×10–6 mbar l/s.