Imprimer

Hygromycine B 50 mg/ml dans un tampon phosphate salin (de Streptomyces hygroscopicus) , 90% (par HPLC), filtré stéril

Marques : Enzo Life Sciences
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ALEXALX380306G001EA 211 EUR
ALEXALX380306G001
Hygromycine B 50 mg/ml dans un tampon phosphate salin (de Streptomyces hygroscopicus) , 90% (par HPLC), filtré stéril
Hygromycine B

Antibiotic. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent.


  • Protein synthesis inhibitor


Liquid. In PBS


Cdt: Shipping temperature: Ambient

Formule: C₂₀H₃₇N₃O₁₃
Poids moléculaire: 527,5 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD06795479
Numéro CAS: 31282-04-9
https://fr.vwr.com/stibo/web/std.lang.all/05/67/45390567.jpg

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR