Hygromycine B 100 mg/ml dans l'eau (de Streptomyces hygroscopicus) , 90% (par HPLC), filtré stéril
Marques : Enzo Life SciencesTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Antibiotic. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent.
- Protein synthesis inhibitor
Liquid. In deionized water
Formule:
C₂₀H₃₇N₃O₁₃ Poids moléculaire: 527,5 g/mol Température de stockage: Réfrigérateur |
Numéro MDL:
MFCD06795479 Numéro CAS: 31282-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...