Anisomycin ≥97% (par CCM)
Marques : Enzo Life SciencesTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Activator of MAPK, Jnk and p38
Isolated from Streptomyces griseolus.
Antibiotic. Activates JNK/SAPKs, p38 and MAP kinases but not ERKs leading to the phosphorylation of S6, histone H3 and HMG-14 and the induction of the immediate-early genes c-fos and c-jun. Induces apoptosis in the human monoblastoid cell line. Used at 300 ng/ml. Inhibits peptidyl transferase at higher concentrations. Used in the eradication of bean mildew. Inhibits other pathogenic fungi in plants.
Formule:
C₁₄H₁₉NO₄ |
Numéro MDL:
MFCD00077650 Numéro CAS: 22862-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...