(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid ≥98%
Marques : Enzo Life SciencesTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
COX inhibitor. NSAID.
Cyclooxygenase (COX) inhibitor. Widely used non-steroidal anti-inflammatory drug.
Formule:
C₁₄H₁₄O₃ Poids moléculaire: 230,26 g/mol |
Numéro MDL:
MFCD00010500 Numéro CAS: 22204-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...