2,2'-Methylenebis(6-tert-butyl-p-cresol) ≥98.5% (par GC)
Marques : Enzo Life SciencesTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A novel inhibitor of Acyl-CoA Cholesterol Acyltransferase (ACAT) produced by Aspergillus fumigatus F93. Also displays antioxidant activity.
Formule:
C₂₃H₃₂O₂ Poids moléculaire: 340,51 g/mol |
Numéro MDL:
MFCD00043641 Numéro CAS: 119-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...