Imprimer
Membrane Protein Extraction Kit
Fournisseur: PromoKine
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
POMOPK-CA577-K268EA 399 EUR
POMOPK-CA577-K268
Membrane Protein Extraction Kit
Protéines extraction Protein Extraction Kits
The Membrane Protein Extraction Kit provides an easy and complete protocol for isolating integral membrane proteins efficiently from cultured mammalian cells.
  • Extract total cellular membrane proteins
  • Purify plasma membrane proteins
  • Procedure takes less than one hour
The Membrane Protein Extraction Kit has been designed to not only extract the total cellular membrane proteins, but also to specifically purify the plasma membrane proteins. The procedure offers consistent yield and high purity (> 90%). Membrane proteins prepared using the kit can be utilized in a variety of applications, such as Western blotting, 2-D gels, and enzyme analyses, etc. The entire procedure takes less than one hour.