Imprimer
Graduated pipettes, BLAUBRAND®, type 1, Class AS, partial delivery (individual certificate)
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BLAUBRAND®
BRND927731EA 67.5 EUR
BRND927731 BRND927722 BRND927727 BRND927729 BRND927725
Graduated pipettes, BLAUBRAND®, type 1, Class AS, partial delivery (individual certificate)
Pipettes Pipettes graduées
AR-Glas®.
  • Calibrated to deliver (TD, Ex); type 1: Zero on top
  • Marks and inscriptions in high contrast blue enamel
  • Individual certificate
  • Only pipettes marked (*) are suitable for cotton plugs
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Certifications: DIN EN ISO 835. DE-M marking.