Imprimer
Waste bins, Hospisafe
Fournisseur: Hospidex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HUARBG50LATEGHEA 16.2 EUR
HUARBG50LATEGH HUARBG60LATEGH HUARBG30LATEGH
Waste bins, Hospisafe
Poubelles Conteneurs de déchets
The containers are made of pure PP, when burned no pollutants are released.
  • Leakproof and gastight after closing
  • Very good chemical resistance
Certifications: UN approved and conforms with NFX 30511, ADR and NFX 30505.