Imprimer
Salt tester, Eutech EcoTestr Salt
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Eutech EcoTestr
EUTEECOSALTTESTEA 80.3 EUR
EUTEECOSALTTEST
Salt tester, Eutech EcoTestr Salt
Compteurs électrochimiques Compteurs de salinité
This waterproof and accurate tester with stainless pin sensor is ideal for basic salinity measurements in aquaculture, spas, pools, simple lab work and other water/wastewater applications.
  • Manual calibration option for better accuracy with near-to-sample calibrations
  • Hold function freezes reading temporarily for ease of recording
  • IP67 waterproof – tester floats on water for easy retrieval
  • Click-lock battery compartment requires no tools for battery replacements
  • Transparent protective cap doubles up as a container for samples or calibration solution
  • Large, upright LCD with parameter display and battery level indicator for easy reading