Imprimer
Acrylamine/Bis-Acrylamine, Acryl/Bis™
Fournisseur: VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M157-500MLEA 0 EUR
M157-500ML
Acrylamine/Bis-Acrylamine, Acryl/Bis™
Gels d'électrophorèse Agarose et acrylamide
A 40% (w/v) solution of ultra pure acrylamide (38,96%) and bis-acrylamide (1,04%) at a final ratio of 37.5:1.
  • Ultra-pure acrylamide powders (>99,9%)
  • High solubility, producing haze-free solutions
  • Liquid stable solutions eliminate handling hazardous powdered acrylamide
  • Reproducible and consistent results
VWR offers an extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions and ready-to-use solutions for customised polyacrylamide gel electrophoresis of proteins. Ultra-pure (>99,9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimise the handling of neurotoxic acrylamide.