Imprimer
Melting Point Standard, Stearic acid, 69 - 71°C, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SUPL77402-1GEA 24.8 EUR
SUPL77402-1G
Melting Point Standard, Stearic acid, 69 - 71°C, Supelco®
Melting point standard with applications in the cleaning products, cosmetics, flavors and fragrances, food and beverages and personal care sectors.
Attention: Limited shelf life, expiry date on the label