Imprimer
Stopcocks, three-way, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S7521-10EAEA 111 EUR
S7521-10EA
Stopcocks, three-way, Supelco®
Robinets d'arrêt
Made of nylon.
  • Good chemical resistance
  • Good temperature resistance
Features three-way control with female luer-lock inlet and male luer-lock outlet ports.