Imprimer
Peqlab peqGOLD, DNA Ladder, 100 bp Plus
Fournisseur: VWR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Peqlab peqGOLD
25-2020EA 73.6 EUR
25-2020
Peqlab peqGOLD, DNA Ladder, 100 bp Plus
Réactifs d'électrophorèse Marqueurs de poids moléculaires
peqGOLD DNA ladder 100 bp plus is ideal for size determination in the range of 100 to 3000 base pairs. There are no unspecific bands besides the fragments described. 1000 and 500 bp bands have a higher DNA content and serve as a size reference.
  • Composed of 14 chromatography-purified individual DNA fragments: 3000, 2000, 1500, 1200, 1000, 900, 800, 700, 600, 500, 400, 300, 200 and 100 base pairs
Informations de livraison: Product comprises of 100 µl (50 µg) DNA ladder and 1 ml 6X loading buffer (10 mM Tris-HCl, pH 7,6; 0,03% bromophenol blue; 0,03% xylene cyanol; 60% glycerol and 60 mM EDTA)
Attention: The marker has to be dephosphorylated before being used in radioactive assays.