Imprimer
siRNA transfection reagent, si3D-Fect™
Fournisseur: OZ Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
si3D-Fect™
OZBISTF40250EA 118.25 EUR
OZBISTF40250 OZBISTF40500 OZBISTF41000
siRNA transfection reagent, si3D-Fect™
Réactifs de transfection
si3D-Fect™ is a 3D transfection reagent specifically designed and developed for silencing gene expression in cells cultured on 3D scaffolds.
  • Highly efficient for gene silencing in 3D matrices
  • Ideal for any 3D scaffolds (sponges, matrices, inserts)
  • Dedicated to short nucleic acid sequences such as siRNA and miRNA
  • Universal - suitable for primary cells and cell lines
  • Serum compatible
  • Shipped at room temperature
si3D-Fect™ reagent associated with 3D matrices allows numerous cell transfections in order to study tissue engineering, tissue regeneration, tumour invasion, neural differentiation, cellular polarisation, tissue formation, colonisation and neurite growth.
Attention: Store at 4 °C.