Imprimer
Magnetic sets
Fournisseur: OZ Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
OZBIIV-MAG2EA 96.75 EUR
OZBIIV-MAG2 OZBIIV-MAG1 OZBIIV-MAG3
Magnetic sets
Aimants
These magnetic sets have been developped for efficient in vivo transfection or transduction with in vivo Magnetofection technology.
  • Can be easily cleaned and decontaminated with 70% ethanol, or by dry or wet sterilisation
  • Can be used at room temperature, 37 °C, +4 °C, etc.
  • Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targetd site
  • Solid, completely reusable, it is a one-time buy
Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targeted site.