Imprimer
Transfection reagent, PolyMag™ CRISPR
Fournisseur: OZ Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PolyMag™
OZBIPNC40200EA 251.34 EUR
OZBIPNC40200 OZBIKPC40100 OZBIPNC41000
Transfection reagent, PolyMag™ CRISPR
Réactifs de transfection
PolyMag CRISPR is a magnet-assisted transfection reagent optimised to deliver high levels of plasmid DNA and/or mRNA expressing Cas9 and guide RNA (gRNA).
  • High efficiency delivery of CRISPR/Cas9 system, even on primary and hard to transfect cells
  • Ideal for co-transfection of pDNA/pDNA, pDNA/gRNA, pDNA/mRNA
  • Optimised for large plasmid DNA containing Cas9 and guide RNA
  • Low cell toxicity
PolyMag CRISPR reagent is based on Magnetofection™ technology. The nanoparticles are composed of iron oxide magnetic core coated with cationic molecules. For gene editing experiments, this reagent provides high transfection efficiency without toxicity.
Store at +4 °C
Informations de commande: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.