Storage Boxes for Vials
Supplier: Hycultec
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SANB9111-1PP
Storage Boxes for Vials
Storage box made of polypropylene for 96 vials of 4 ml.
  • Very good temperature resistance
  • Very good chemical resistance