Imprimer
Cell culture media, Dulbecco's Modified Eagles Medium (DMEM)
Fournisseur: United States Biological
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
USBID9812-05-50LEA 377 EUR
USBID9812-05-50L
Cell culture media, Dulbecco's Modified Eagles Medium (DMEM)
Dulbecco's MEM (DMEM) is the most widely used modification of BME. DMEM was originally developed for the culture of mice embryonic cells. Today, it finds broad application in serum-free culture of normal and transformed mouse and chicken cells.