Imprimer
Guarded flow meters, Riteflow®, SP Bel-Art
Fournisseur: Bel-Art
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BELAH404030235EA 1130 EUR
BELAH404030235
Guarded flow meters, Riteflow®, SP Bel-Art
Débitmètres
These flow meters has a plastic safety guard over the flowtube.
  • Heavy-walled shock-resistant borosilicate glass
  • Fused and enclosed ceramic scales
  • Uniformly tapered bores with internal rib guides
  • Floats are retained by Teflon® plugs
Used for flow measurements in the lab, process flows in pilot plants, metering carrier gases in chromatography, indicating and controlling gases in manufacturing processes, and for indicating flow rates in pumping or flow systems.