Imprimer
Pellet press, PP 35
Fournisseur: Retsch
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RETS207550001EA 23550 EUR
RETS207550001
Pellet press, PP 35
The PP 35 is a small benchtop pellet press with high pressure forces up to 35 t.
  • Produces strong pellets with a smooth surface
  • Individual pressure force adjustment up to 35 tons
  • Pressing in steel rings, aluminium cups and free pressing tools for various diameters
  • 10 SOPs can be defined and stored for routine applications
  • Comfortable parameter setting via display
  • Automatic pressure force control
Ideal for the preparation of pellets for spectral analysis. Fields of application include: Chemistry / plastics, construction materials, environment / recycling, geology / metallurgy, glass / ceramics.