Imprimer
Vacuum pump, VACSTAR control
Fournisseur: IKA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IKAA20019065EA 5420 EUR
IKAA20019065
Vacuum pump, VACSTAR control
Pompe Pompes à vide pour évaporateurs rotatifs
VACSTAR control is the ideal system with its integrated vacuum controller and removable display. The controller offers a suitable mode for the desired process: from automatic to manual operation, right through to pumping or an evacuation and cleaning function.
  • High-performance suction with its 22 l/min at a minimum of 2 mbar
  • Integrated vacuum controller with removable WiCo
Includes a downstream vacuum safety emission condenser (VSE1). This reliably protects laboratory employees and the environment from harmful exhaust air.
Suction levels: 4, cylinder: 4, connection Ø suction side: 8 mm, input pressure: 2 to 1030 mbar, speed control: turning knob, display: TFT, vacuum sensor type: ceramic Al₂O₃, measurement range (absolute): 1 to 1030 mbar, resolution pressure and measurement uncertainty: 1 mbar, noise at low pressure: 54 dB(A), material in contact with medium: Al₂O₃, PTFE, FPM, PPS, housing material: alu-cast coating/thermoplastic polymer, permissible relative humidity: 80%.
Certifications: Protection class according to DIN EN 60529; IP 20.