Imprimer
Trousers, Crafts, NYC-2018, Cordura®
Fournisseur: Fristads Kansas Group
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Cordura®
KWIN113236940D108EA 86.6 EUR
KWIN113236940D108 KWIN113236940D104 113-8532 113-8533 KWIN113236940D120 113-8530 113-8531 KWIN113236940D100 113-8518 113-8519 KWIN113236940D92 113-8516 113-8517 113-8514 113-8515 KWIN113236940D96 KWIN113236940D116 KWIN113236940D112 113-8521 113-8522 113-8520 113-8529 113-8527 113-8528 113-8525 113-8526 113-8523 113-8524
Trousers, Crafts, NYC-2018, Cordura®
Tenue de travail
Craftsman trousers in strong and light weight ripstop material, with ventilation panels at back and behind knees for increased comfort when working in warm environments.
  • Dynamic, made of 50% cotton and 50% polyamide
  • Maximum washing temperature 85 °C, normal process
  • Tumble drying possible, low temperature, exhaust temperature maximum of 60 °C
  • Iron at maximum sole-plate temperature of 150 °C
Mesh ventilation at back yoke and knee, two loose-hanging CORDURA® reinforced outside pockets, one with zip pocket, one with three smaller pockets and tool loop. D-ring on front belt loop and at back yoke two front pockets, one with phone pocket, two slanted back pockets, wide belt loop at mid back, double reinforced crotch seam. CORDURA® reinforced folding rule pocket with pen holder, button and loop for sheath knife.
Multifunction leg pocket with phone pocket, ID-card pocket and large inside pocket with snap fastening. CORDURA® reinforced knee pockets with inside opening. Height adjustment for knee pads in knee pocket, CORDURA® reinforced leg ends adjustable with snap fastenings, Pockets reinforced with metallic rivets at stress points.
Weight: 240 g/m²
Certifications: EN 14404; with kneepads 124292 / OEKO-TEX®
Attention: Do not bleach or dry clean.