These fume hoods are intended to be connected to an exhausting system for the discharge of toxic and harmful vapours to the outside of the building.
- Steel construction with white epoxy paint RAL 9010 which is highly resistant to chemicals
- Ergonomic hand traps for safe and easy handling in the enclosure
- Hands passage or trapezoidal opening
- Ideal for storage of flammables, corrosives and toxics
Delivered ready to use, no assembly required and to be connected only to the electrical network and to an exhausting system. Transparent glass walls, made of PMMA, on all sides offer optimal illumination on the work top and immediate visualisation of the products handled. Lids allow the introduction of cables for power supply of equipment and fluids inside the ductless fume hood.