Imprimer
Bundle SpectraMax® iD3 + SpectraMax® DuoLuc™ Reporter Assay + SpectraMax® Glo Steady-Luc™ Reporter Assay
Fournisseur: Molecular Devices
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MLDVQ-00108012EA 0 EUR
MLDVQ-00108012
Bundle SpectraMax® iD3 + SpectraMax® DuoLuc™ Reporter Assay + SpectraMax® Glo Steady-Luc™ Reporter Assay
Save on Luciferase Assay solutions!
Get a SpectraMax® iD3 with FREE injectors and FREE Glo Steady LUC reporter assay kits!
Offering absorbance, fluorescence and luminescence read modes, the SpectraMax iD3 multi-mode microplate reader is the complete solution for all your research needs, including Luciferase assays.
  • The SpectraMax® iD3 allows the creation of user-specific custom protocols and personalised workflows, the ability to push your data to a remote workstation, and an intuitive, easy to use interface
  • Optimised kits for use with our SpectraMax® readers: GLO Steady-Luc™ Reporter Assay kit provides an extended signal time window enabling high throughput reads across multiple plates; DuoLuc™ Reporter Assay kit enables highly sensitive quantitation of both firefly and Renilla luciferases in mammalian cells, and increases the accuracy of results by normalising reporter expression to a control
  • Injectors allow equal mixing across the plate during high precision experiments with minimal reagent loss