Imprimer
Laboratory bottles, wide neck, without screw cap, with screw thread, DURAN® protect GLS 80®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SCOT218656909EA 436 EUR
SCOT218656909
Laboratory bottles, wide neck, without screw cap, with screw thread, DURAN® protect GLS 80®
Flacons Flacons en verre
DURAN®, PROTECT coated borosilicate glass 3.3, clear, without screw caps and pouring rings.
  • Scratch, leak and shatter resistant, highly transparent
  • Temperature resistance from −30 to +135 °C
  • Suitable for use in microwaves
  • Glass Type I according to USP, EP and JP
Ideal for handling particularly valuable or dangerous materials. The plastic coating (PU) provides effective protection during routine laboratory work and retains the contents if the glass becomes damaged. Round, with graduations and specially developed GLS 80® thread. Opening and closing the bottle is easy with the quick closure mechanism. The 80 mm wide outer diameter of the bottle neck enables bottles to be filled conveniently and safely with powders, pastes, granulates and liquids. The large opening can accommodate even large volume funnels and gives easy access to scoops or spatulas.
With printed Retrace Code (batch identification); batch certificate available.