Imprimer
Orange filter for myECL™ imager
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Pierce
PIER62238EA 978 EUR
PIER62238
Orange filter for myECL™ imager
Systèmes de photodocumentation
Thermo Scientific™ 565 to 620 nm bandpass filter is a threaded, 50 mm diameter glass filter. This product is a replacement for the pre-installed orange filter in the myECL™ Imager, and is used for detection of colorimetric protein stains (silver, coomassie) and wavelengths of common fluorescent nucleic acid gel stains (e.g., ethidium bromide) and protein stains, such as SYPRO® Orange Protein Gel Stain.