Imprimer
Buccal collection kit, Whatman™ EasiCollect™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WHATWB120237EA 378 EUR
WHATWB120237
Buccal collection kit, Whatman™ EasiCollect™
EasiCollect™ is designed for the collection of buccal cells plus the protection and isolation of DNA using the Whatman™ FTA™ technology.
  • Designed to facilitate collection, transport, and archiving of buccal and saliva samples
  • Kit includes an Indicating FTA™ Card that changes colour from pink to white upon sample addition for easy visualisation
  • Allows conventional and direct STR amplification chemistries for easy implementation into workflows
  • Compatible with Hamilton Robotics’ easyPunch™ automation and Cytiva UniCore Punch for manual processing
  • Enables long-term archiving of samples at room temperature
Cells are captured on the foam tip of the EasiCollect™ by swabbing the oral mucosa and subsequently transferred to the integral Indicating FTA™ Card. Upon contact, the proprietary chemical formulation lyses cells, denatures proteins and promotes the binding of DNA to the card matrix. The pink indicating dye on the card will turn white to mark the location of the sample. The card chosen for the EasiCollect™ is compatible with manual and automated punching systems to meet laboratory sample throughput requirements.
Certifications: Manufactured to ISO9001:2008 standards.
Informations de livraison: Kit contains an EasiCollect™ device, peelable barcodes, gloves, a Multi-Barrier Pouch, a mailing envelope, tamper-proof tape for the mailing envelope, desiccant, and a clear outer bag labeled with code number and expiration date. EasiCollect™ device also supplied separately.