Imprimer
Alkali metal mix for ICP, 5 elements, TraceCERT®, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TraceCERT®
96441-100MLEA 136 EUR
96441-100ML
Alkali metal mix for ICP, 5 elements, TraceCERT®, Supelco®
Étalons Étalons cationiques Standards multi-éléments
100 mg/L each: Li, Na, K, Rb and Cs in 2% nitric acid
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034