Imprimer
Chemical resistant gloves, Tychem® PV350
Fournisseur: DuPont
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Tychem®
112-5498EA 0 EUR
112-5498
Chemical resistant gloves, Tychem® PV350
Gants
These cotton/jersey gloves with PVC coating are extremely durable and perfect for defensive protection against chemicals, oil and grease.
  • Abrasion resistance
  • Versatility
  • Rough grip
Ideal for all general-purpose industrial applications.
Certifications: EN 388:2016 (3111X), EN ISO 374-1:2016 Type B (AKLMP), EN ISO 374-5:2016 (VIRUS); Cat. III