Imprimer
Chemical resistant gloves, Tychem® NT480
Fournisseur: DuPont
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Tychem®
112-5492EA 0 EUR
112-5492 112-5493 112-5494 112-5495 112-5496 112-5497
Chemical resistant gloves, Tychem® NT480
Gants
These lightweight flock-lined gloves with 100% nitrile coating are designed to last and reduce fatigue. They are resistant to a range of solvents, animal fats and other chemicals.
  • Unsupported
  • Bisque finish allows secure handling in wet and dry conditions
  • Wrist protection
  • Chlorinated
  • Flock-lined for enhanced comfort
Ideal for working safely in damp or greasy environments.
Certifications: EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016 (VIRUS), EN 388:2016 (2001X), Cat. III.
Food contact approved.