Imprimer
Dynamic climate chambers, MKF series
Fournisseur: Binder
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BIND9020-0208EA 0 EUR
BIND9020-0208
Dynamic climate chambers, MKF series
MKF series environmental simulation chamber is ideally suited for any cold and heat testing based on current temperature. The comprehensive standard equipment for this environmental simulation chamber ensures ease of operation.
  • Programmable condensation protection for test material
  • Heated viewing window with LED interior lighting
  • Integrated water-storage tank, 20 litres
  • APT.line™ preheating chamber technology
  • CFC-free refrigerant R-452A
  • Four zero-voltage relay contacts
These chambers have intuitive touchscreen controller with time-segment and real-time programming, and internal data logger, measured values can be read out in open format via USB.
Certifications: They meet DIN EN 60068-2-30: 2006-06, DIN EN 60068-2-38: 2010-06, MIL STD 810:2000-01, DIN EN 60068-2-66 standards.
Informations de livraison: They are delivered with four zero-voltage relay contacts and one stainless steel rack.