Imprimer
Pesticide reference standard, Aclonifen, PESTANAL®, analytical standard, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PESTANAL®
36792-50MG-REA 59.4 EUR
36792-50MG-R
Pesticide reference standard, Aclonifen, PESTANAL®, analytical standard, Supelco®
Étalons Étalons organiques
Aclonifen is a diphenyl-ether herbicide, which can be used for weed control management, in specific for sunflowers. Its mode of action involves, the inhibition of carotenoid synthesis and suppression of protoporphyrinogen oxidase enzyme involved in the chlorophyll synthesis pathway.