Imprimer
Pesticide reference standard, Oxyfluorfen, PESTANAL®, analytical standard, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PESTANAL®
35031-100MGEA 105 EUR
35031-100MG
Pesticide reference standard, Oxyfluorfen, PESTANAL®, analytical standard, Supelco®
Étalons Étalons organiques
Oxyfluorfen is a diphenyl-ether herbicide, which can be commonly used in vineyards for weed control management. It can block the chlorophyll biosynthesis in plants by inhibiting protoporphyrinogen oxidase enzyme.