Imprimer
Safety goggle, 5X9
Fournisseur: Univet
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
UNVT5X9.20.00.00EA 35.2 EUR
UNVT5X9.20.00.00
Safety goggle, 5X9
Lunettes de protection
The two-lens structure of the model 5X9 permits to provide the device with alternative materials to polycarbonate lenses, offering protection to a greater number of professional fields.
  • Integrated side shields with ventilation system
  • Has adjustable temple, elastic band and soft rubber gasket
  • SoftPad technology
The integrated flip-up perfectly combines with the glasses forming a unique modular system for maximum flexibility of use, made possible by the wide range of filters.
Weight: 45 g