Imprimer
Goggles, ultravision wide-vision gas-tight
Fournisseur: Uvex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
uvex ultravision
UVEX9301.613EA 32.7 EUR
UVEX9301.613
Goggles, ultravision wide-vision gas-tight
Lunettes de protection
Classic, ergonomic wide-vision goggle with unrestricted side visibility.
  • Anti-fog coating on the inside, extremely resistant to scratches and chemicals on the outside
  • Gas-tight version without a ventilation system
  • Large goggle body, can also comfortably be worn as overspecs
  • Yellow frame
  • Clear PC lens
The uvex ultravision features a comfortable and ergonomic design and offers unrestricted side visibility. This flexible wide-vision goggle is also suitable for use as overspecs. The uvex supravision excellence coating provides anti-fog protection on the inside and makes the outside extremely resistant to chemicals and scratches. Lenses can be replaced.