Imprimer
Organic reference standard, Octyl gallate, analytical standard, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06717-500MGEA 76.5 EUR
06717-500MG
Organic reference standard, Octyl gallate, analytical standard, Supelco®
Étalons Étalons organiques
Octyl gallate is an ester of gallic acid which is generally used as an antioxidant in food and pharmaceutical industries. It can act against the oxidative damage caused by reactive oxygen species (ROS) in the form of hydroxyl radicals or hydrogen peroxides, and reactive sulphur species (RSS).
Octyl gallate can be used as a reference standard for the determination of octyl gallate in edible vegetable oil by high performance liquid chromatography/time-of-flight mass spectrometry (HPLC/TOF-MS) as well as alternating penalty trilinear decomposition (APTLD) combined with HPLC-diode array detection (DAD) dry foods by reversed-phase HPLC with photodiode array (PDA) detection.
Cdt: Bottomless glass bottle. Contents are inside inserted fused cone.