Imprimer
Nitrogen generator, Infinity XE 5020
Fournisseur: Peak Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PEAK3300573EA 17282 EUR
PEAK3300573
Nitrogen generator, Infinity XE 5020
Générateurs de gaz Générateurs d'azote
Producing between 20 and 120 L/min of high purity nitrogen gas, the Infinity XE 5020 membrane nitrogen generator can comfortably supply multiple LC-MS instruments with LC-MS grade nitrogen.
  • Variable flow rate
  • Purity from 95 to 99,5% (flow rate dependent)
  • Tried and tested membrane technology for high purity gas
  • Small footprint
Dew point: –40 °C
Inlet pressure: 8,6 bar
Outlet pressure: 6,8 bar
Certifications: Conforms with: Low Voltage Directive 2014/35/EU
EN 61010-1: 2010 Electrical Equipment for measurement, control and laboratory use.
Electromagnetic Compatibility Directive 2014/30/EU
EN 61326-1: 2013 Electrical Equipment for measurement, control and laboratory use.
ISO8753-1:2010 Class 1.4.1.