Imprimer
Multi-element Anion HPCE standard solution, analytical standard, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
29235-50MLEA 50.7 EUR
29235-50ML
Multi-element Anion HPCE standard solution, analytical standard, Supelco®
Étalons Étalons anioniques Standards anioniques à plusieurs éléments
Standard employed in the assay of anions by capillary electrophoresis.
  • Prepared with high purity salts and water
Bromide, 100 ppm
Chloride, 100 ppm
Fluoride, 100 ppm
Nitrate, 100 ppm
Phosphate, 100 ppm
Sulphate, 100 ppm