Imprimer
Tartrate standard for IC, certified reference material, TraceCERT®, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SUPL43484-100MLEA 39.7 EUR
SUPL43484-100ML
Tartrate standard for IC, certified reference material, TraceCERT®, Supelco®
Prepared with tartaric acid and water; filtered through a 0,2 μm membrane filter and stabilised with sodium azide.
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.