Imprimer
Wall-mounted hose racks, stainless steel
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VWRI734-3222EA 205 EUR
VWRI734-3222 VWRI734-3223 VWRI734-3220 VWRI734-3221 VWRI734-3219 VWRI734-3218
Wall-mounted hose racks, stainless steel
The arch and slot style hose racks accommodate a variety of hoses, and are available in standard or mini sizes. These wall-mounted racks keep various size hoses organized and off the floor.
  • Extends hose working life and ensures 100% drainage
  • Compact designs ideal for use in any cleanroom environment
  • Eliminates clutter and enhances safety
  • Conveniently stores sanitary gaskets, clamps, and adaptors in one location (not included on Arch Rack Mini)
  • Constructed from 304 electropolished stainless steel
  • Ideal for biopharmaceutical cleanroom applications
Three unique wall-mounted hose rack styles are available to choose from, including slot, arch, and mini-arch shaped designs.
Certifications: Meets FDA and 3A regulations for hose storage, arch and slot style racks suits cGMP.
Informations de livraison: Supplied with mounting hardware.