Imprimer
VWR® Super Compact, Glass Columns
Fournisseur: VWR, part of Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
554-4093EA 659 EUR
VWRI554-4093 VWRI554-4092 VWRI554-4091 VWRI554-4090 VWRI554-4112 VWRI554-4123 VWRI554-4122 VWRI554-4121 VWRI554-4120 VWRI554-4119 VWRI554-4118 VWRI554-4117 VWRI554-4116 VWRI554-4115 VWRI554-4114 VWRI554-4125 VWRI554-4124 VWRI554-4113
VWR® Super Compact, Glass Columns
Super Compact glass columns are used for analytical chromatography.
  • Stable in high pressure
  • Easy handling
  • Great flexibility of the system
  • Wide product range makes it an efficient system for the chromatographic analysis and chromatographic development of methods
Used in areas such as analytical laboratory and in development of methods for research.
Glass helps in the control of the status of the column during filling, by the chromatography and by the regeneration and the storage.
Piston with the known lip seas unique design guarantees zero dead volume sample injection by sealing directly on the gel bed.
Informations de livraison: Supplied with filter-set, connection-set and filter-adjustment-stick.