Imprimer
Avantor® Hichrom, Female to Female Luer Adapter
Fournisseur: VWR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Avantor® Hichrom
HICHHI-314EA 33.2 EUR
HICHHI-314
Avantor® Hichrom, Female to Female Luer Adapter
Raccords et connecteurs
This adapter provides us the opportunity to adapt female luer to female luer connection to a threaded connection.