Imprimer
Face mask with ear loop
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HOITA-31BR-BAUKEA 5.8 EUR
HOITA-31BR-BAUK
Face mask with ear loop
These are designed with excellent bacterial and particle filtration combined.
  • Easy breathing and non-irritant
  • Made of PP
Present with adjustable clamp for better fit.
Bacterial filter efficiency: 99%
Cdt: 50 pieces/box, 20 boxes/case.