Imprimer
Fluoride Activity Standard Solution for Fluoride Electrode, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SUPL443174-475MLEA 113 EUR
SUPL443174-475ML
Fluoride Activity Standard Solution for Fluoride Electrode, Supelco®
Activity standard solution consists of sodium fluoride solution. 100 ppm as fluoride.
It may be used as calibration standard in determination of fluoride in bones using fluoride electrode.
100 ppm = 100 mg/L (as F).