Imprimer
Filter bags, BagFilter® Pipet & Roll
Fournisseur: Interscience
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BagFilter®
INSC111720EA 327 EUR
INSC111720 INSC111700 INSC111710
Filter bags, BagFilter® Pipet & Roll
Sachets Sachets d'homogénéisation
The BagFilter® Pipet & Roll bags are rigid and transparent blender bags with a lateral non-woven filter for all microbiological analyses. They are ideal for homogenising fibrous samples and compatible with any blender.
  • Multilayer®: reinforced multicoated complex
  • Filter porosity: < 250 microns
  • Resistant to freezing and high temperatures (from –40 °C to 80 °C / –40 °F to 176 °F)
  • Gamma ray treated: Gamma 5 to 12 kGy, with certificate
  • Average shelf-life: 5 years
  • Storage: in a temperate and dry place, away from humidity and light
The reclosable sticker allows easy pipetting with a micropipette cone. Cross contamination risks are reduced with pipet cones for pathogens pre-enrichment bags. The lower pipetting height reduces risks of MSD (Musculoskeletal risks). A 100% metal free sticker built-in closure allows a water-tight storage of the bag, simply roll the top of the bag 3 times. No contact between the sample and the blender during homogenisation of the sample. Filtration is instantaneous without cross-contamination.
BagFilter® Pipet & Roll: With built-in closure sticker and reclosable easy pipetting sticker. Compatible with BagRack® Slide.
BagFilter® Pipet: With reclosable easy pipetting sticker. Compatible with BagRack® Slide with BagClip®.
BagFilter® Roll: With built-in closure sticker. Compatible with BagRack® Slide.
Certifications: Approved for food contact: Regulation (EC) No 1935/2004. In compliance with ISO 7218, ISO 6887-1 and FDA BAM (Bacteriological Analytical Manual).