This digital magnifier has been designed for applications where viewing, image capture, reporting and documentation is needed.
- Fast and accurate image capture
- 4,3" colour display with dimension grid
- Virtual calliper display with movable filar lines
- Live view via mini-HDMI output
- Controllable LED illumination (for 4 level settings: 100%, 75%, 50%, 25%) optimised for magnified imaging
- Stores up to 20000 images
- Image security (no connection to Wi-Fi)
- Secure grip for left- and right-hand use
Ideal for roaming inspectors, documenting faults, or for inspecting immobile subjects. It can be used in hard to reach areas, such as high up inside large machines as well as remote locations to access wild flowers and insects.
View modes:
- Full colour
- Black and white
- Inverted black and white
- Grey scale
Dimension grids:
- Fixed 1 mm² grid lines
- Simple X,Y on-screen dimensioning
- Virtual caliper
Image capture resolution: 1280×720, JPEG
Video output: Mini-HDMI TV-out
Internal memory: 4 GB
Connections: Micro USB connection for image download and charging and Mini-HDMI for monitor
Battery: Lithium ion, 6 hour continuous operation time
Weight: 205 g
Dimensions (L×H×D): 283×88×22 mm
Informations de livraison: Supplied ready to use with soft case, USB cable, mini-HDMI-HDMI cable, TV out cable, Lanyard, cleaning cloth and user guide.