Imprimer
ESD safety shoes, lace-up, uvex 1 G2, 6846
Fournisseur: Uvex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
uvex 1 G2 uvex 1
UVEX6846.9/36EA 114 EUR
UVEX6846.9/36 UVEX6846.9/35 UVEX6846.7/51 UVEX6846.7/52 UVEX6846.7/50 UVEX6846.9/39 UVEX6846.9/38 UVEX6846.9/37 UVEX6846.9/47 UVEX6846.8/35 UVEX6846.9/46 UVEX6846.9/45 UVEX6846.9/44 UVEX6846.9/43 UVEX6846.9/42 UVEX6846.9/41 UVEX6846.9/40 UVEX6846.0/39 UVEX6846.0/38 UVEX6846.0/37 UVEX6846.0/36 UVEX6846.0/35 UVEX6846.8/39 UVEX6846.8/38 UVEX6846.9/49 UVEX6846.8/37 UVEX6846.9/48 UVEX6846.8/36 UVEX6846.8/46 UVEX6846.8/45 UVEX6846.8/44 UVEX6846.8/43 UVEX6846.8/42 UVEX6846.8/41 UVEX6846.9/52 UVEX6846.8/40 UVEX6846.9/51 UVEX6846.9/50 UVEX6846.0/49 UVEX6846.0/48 UVEX6846.0/47 UVEX6846.0/42 UVEX6846.0/41 UVEX6846.7/39 UVEX6846.0/40 UVEX6846.0/46 UVEX6846.7/37 UVEX6846.8/49 UVEX6846.0/45 UVEX6846.7/38 UVEX6846.0/44 UVEX6846.7/35 UVEX6846.8/48 UVEX6846.8/47 UVEX6846.0/43 UVEX6846.7/36 UVEX6846.7/44 UVEX6846.7/45 UVEX6846.7/42 UVEX6846.7/43 UVEX6846.7/40 UVEX6846.8/52 UVEX6846.7/41 UVEX6846.8/51 UVEX6846.8/50 UVEX6846.0/52 UVEX6846.0/51 UVEX6846.0/50 UVEX6846.7/48 UVEX6846.7/49 UVEX6846.7/46 UVEX6846.7/47
ESD safety shoes, lace-up, uvex 1 G2, 6846
Chaussures et bottes
Ultra-lightweight, perforated S1 safety shoes are available in multiple widths.
  • Made from synthetic materials, making it suitable for people allergic to chrome
  • Breathable micro velour upper material
  • uvex i-PUREnrj with optimum shock-absorption and energy return
  • Meets the ESD specification with volume resistance under 35 megaohms
  • Elastic laces for quick, individual adjustment (normal laces also provided)
The new, innovative uvex i-PUREnrj sole technology offers the highest level of shock-absorbing properties at the forefoot and at the heel, resulting in excellent rebound over the entire midsole and optimal stability thanks to the foam heel basket. A new, wider, 100% metal-free uvex xenova® safety toe cap provides more toe room and an optimum fit. This toe cap offers compact design, anatomical shape, good lateral stability, and no thermal conductivity. All sole materials are free of silicone, plasticisers, and other paint wetting impairment substances.
Colour: Black/orange
Certifications: EN 61340 (ESD); EN ISO 20345:2011 S1 SRC certified.