Imprimer
Nitrile gloves, Showa 7540
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
112-5904EA 69.3 EUR
112-5904
Nitrile gloves, Showa 7540
This thin nitrile disposable glove offers high-performance protection against chemical penetration and splashing.
  • Twice chlorinated to reduce tackiness and create a soft, second-skin feel
  • Latex, silicone and powder-free composition reduces risk of allergies and irritation
  • Textured finish on the fingertips enhances grip when handling small components
Suitable applications include: Intricate parts handling, light assembly of oil-coated pieces, painting & spray workshops, HoReCa, emergency services, medical, cytostatics, laboratory, pharma & analysis, pharmaceuticals and API.
Certifications: EN 455, EN 374-1:2016 Type C (JK), EN 374-5:2016, EN 388 (1000X); Cat. III
Food contact approved
Cdt: 100 gloves per dispenser.