Imprimer
Gasprofi 1 SCS micro school edition
Fournisseur: WLD-TEC GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WART6.007.000EA 641 EUR
WART6.007.000
Gasprofi 1 SCS micro school edition
The safety burner gasprofi 1 micro school edition offers the highest safety standards when handling the flame. Suitable for connection to the fully automatic inoculation loop carousel autoloop Pro.
  • Safe gas burner for flaming and heating combines all functional advantages
  • Flame is automatically reignited
  • Direct switching on and off of the flame by pressing a button without any matches
  • Can also be operated using a foot pedal or an external IR motion sensor
The flame can be precisely adjusted with the dual button for gas and air adjustment. All external components are made of stainless steel and guarantee the highest degree of sterility. Spilled liquids flow out through a channel.
Ideal for connection to natural gas or propane or butane gas.